siRNA Details
Gene Symbol: LOC689065 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109521.1 | NP_001102991.1 | 2 | 388 | ||
XM_006230905.2 | XP_006230967.1 | 3 | 973 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC689065 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109521.1 | NP_001102991.1 | 2 | 388 | ||
XM_006230905.2 | XP_006230967.1 | 3 | 973 |