siRNA Details
| Gene Symbol: Cd300lg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001061779.4 | XP_001061779.2 | 3 | 519 | ||
| XM_003750936.3 | XP_003750984.1 | 3 | 519 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd300lg| Gene Symbol: Cd300lg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001061779.4 | XP_001061779.2 | 3 | 519 | ||
| XM_003750936.3 | XP_003750984.1 | 3 | 519 | |||