siRNA Details
| Gene Symbol: LOC685964 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109489.1 | NP_001102959.1 | 4 | 584 | ||
| XM_006236881.2 | XP_006236943.1 | 4 | 665 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC685964| Gene Symbol: LOC685964 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109489.1 | NP_001102959.1 | 4 | 584 | ||
| XM_006236881.2 | XP_006236943.1 | 4 | 665 | |||