siRNA Details
| Gene Symbol: Eif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105837.1 | NP_001099307.1 | 2 | 283 | ||
| GenBank mRNA | BC159406.1 | - | 2 | 269 | 8361778 | |
| FQ212688.1 | - | 2 | 275 | |||
| FQ213689.1 | - | 2 | 132 | |||
| FQ217644.1 | - | 1 | 41 | |||
| FQ228394.1 | - | 2 | 277 | |||

Eif1
Genomic Map