siRNA Details
| Gene Symbol: Fgf22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130751.1 | NP_570107.1 | 2 | 223 | ||
| GenBank mRNA | AB078902.1 | - | 2 | 223 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgf22| Gene Symbol: Fgf22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130751.1 | NP_570107.1 | 2 | 223 | ||
| GenBank mRNA | AB078902.1 | - | 2 | 223 | ||