siRNA Details
| Gene Symbol: Slc16a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012716.2 | NP_036848.1 | 4 | 1045 | ||
| GenBank mRNA | AJ236865.1 | - | 1 | 65 | ||
| BC078877.1 | - | 4 | 1045 | 7122399 | ||
| D63834.1 | - | 1059 | ||||
| X86216.1 | - | 1065 | ||||

Slc16a1
Genomic Map