siRNA Details
| Gene Symbol: Gucy2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013170.1 | NP_037302.1 | 26 | 3020 | ||
| GenBank mRNA | M55636.1 | - | 26 | 3020 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gucy2c| Gene Symbol: Gucy2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013170.1 | NP_037302.1 | 26 | 3020 | ||
| GenBank mRNA | M55636.1 | - | 26 | 3020 | ||