siRNA Details
| Gene Symbol: Snn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034083.1 | NP_001029255.1 | 2 | 498 | ||
| GenBank mRNA | BC100111.1 | - | 2 | 498 | 7380565 | |
| M81639.1 | - | 2 | 481 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snn| Gene Symbol: Snn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034083.1 | NP_001029255.1 | 2 | 498 | ||
| GenBank mRNA | BC100111.1 | - | 2 | 498 | 7380565 | |
| M81639.1 | - | 2 | 481 | |||