siRNA Details
| Gene Symbol: Cyp2b12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017156.1 | NP_058852.1 | 1465 | |||
| GenBank mRNA | X63545.1 | - | 1465 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp2b12| Gene Symbol: Cyp2b12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017156.1 | NP_058852.1 | 1465 | |||
| GenBank mRNA | X63545.1 | - | 1465 | |||