siRNA Details
| Gene Symbol: Rpl21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | FQ221902.1 | - | 6 | 502 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpl21| Gene Symbol: Rpl21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | FQ221902.1 | - | 6 | 502 | ||