siRNA Details
| Gene Symbol: Htt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024357.3 | NP_077333.2 | 46, 47 | 6390 | ||
| XM_006251230.2 | XP_006251292.1 | 45, 46 | 6328 | |||
| XM_006251232.2 | XP_006251294.1 | 46, 47 | 6376 | |||
| XM_008770274.1 | XP_008768496.1 | 44, 45 | 5842 | |||
| GenBank mRNA | U18650.1 | - | 46, 47 | 6234 | ||

Htt
Genomic Map