siRNA Details
Gene Symbol: Cisd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106385.2 | NP_001099855.1 | 2 | 311 | ||
GenBank mRNA | BC159419.1 | - | 2 | 311 | 8370407 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cisd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106385.2 | NP_001099855.1 | 2 | 311 | ||
GenBank mRNA | BC159419.1 | - | 2 | 311 | 8370407 |