siRNA Details
| Gene Symbol: Nupr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134570.1 | NP_001128042.1 | 2, 3 | 363 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nupr1l| Gene Symbol: Nupr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134570.1 | NP_001128042.1 | 2, 3 | 363 | ||