siRNA Details
| Gene Symbol: Olr960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000589.1 | NP_001000589.1 | 253 | |||
| Gene Symbol: Olr1000 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000077.1 | NP_001000077.1 | 1 | 253 | ||
| Gene Symbol: Olr959 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000486.1 | NP_001000486.1 | 1 | 253 | ||

Olr960
Genomic Map