siRNA Details
Gene Symbol: Olr213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000735.1 | NP_001000735.1 | 1 | 520 |
Gene Symbol: Olr211 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000196.1 | NP_001000196.1 | 1 | 520 |
