siRNA Details
| Gene Symbol: Hdgf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053707.2 | NP_446159.1 | 3 | 548 | ||
| GenBank mRNA | AF389348.1 | - | 3 | 277 | ||
| AF448810.1 | - | 3 | 277 | |||
| BC070943.1 | - | 3 | 548 | 7103046 | ||
| FQ231256.1 | - | 4 | 311 | |||

Hdgf
Genomic Map