siRNA Details
| Gene Symbol: Suclg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053752.2 | NP_446204.2 | 8 | 925 | ||
| XM_008762973.1 | XP_008761195.1 | 7 | 838 | |||
| GenBank mRNA | BC061537.1 | - | 8 | 929 | 6920009 | |
| FQ212668.1 | - | 8 | 922 | |||
| FQ214532.1 | - | 8 | 884 | |||
| FQ214959.1 | - | 8 | 914 | |||
| FQ215028.1 | - | 8 | 883 | |||
| FQ215469.1 | - | 8 | 922 | |||
| FQ216893.1 | - | 8 | 915 | |||
| FQ217244.1 | - | 8 | 939 | |||

Suclg1
Genomic Map