siRNA Details
| Gene Symbol: Ccl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053858.1 | NP_446310.1 | 1, 2 | 81 | ||
| XM_006246966.2 | XP_006247028.1 | 1, 2 | 205 | |||
| GenBank mRNA | EF121996.1 | - | 1, 2 | 81 | ||
| EF121997.1 | - | 1, 2 | 81 | |||
| U06434.1 | - | 1, 2 | 81 | |||

Ccl4
Genomic Map