siRNA Details
| Gene Symbol: Vcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053864.2 | NP_446316.1 | 12 | 1636 | ||
| GenBank mRNA | BC060518.1 | - | 12 | 1636 | 5597843 | |
| U11760.1 | - | 12 | 1668 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vcp| Gene Symbol: Vcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053864.2 | NP_446316.1 | 12 | 1636 | ||
| GenBank mRNA | BC060518.1 | - | 12 | 1636 | 5597843 | |
| U11760.1 | - | 12 | 1668 | |||