siRNA Details
| Gene Symbol: Mark1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053947.1 | NP_446399.1 | 9, 10 | 1062 | ||
| XM_008769816.1 | XP_008768038.1 | 10, 11 | 1066 | |||
| GenBank mRNA | Z83868.1 | - | 9, 10 | 1062 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mark1| Gene Symbol: Mark1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053947.1 | NP_446399.1 | 9, 10 | 1062 | ||
| XM_008769816.1 | XP_008768038.1 | 10, 11 | 1066 | |||
| GenBank mRNA | Z83868.1 | - | 9, 10 | 1062 | ||