siRNA Details
| Gene Symbol: Sh3bp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054011.1 | NP_446463.2 | 8, 9 | 1156 | ||
| XM_008770976.1 | XP_008769198.1 | 6, 7 | 3018 | |||
| GenBank mRNA | AB027562.1 | - | 8, 9 | 1156 | ||
| FQ210473.1 | - | 9, 10 | 1374 | |||

Sh3bp5
Genomic Map