siRNA Details
| Gene Symbol: Oxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001197907.1 | NP_001184836.1 | 9 | 1712 | ||
| NM_057153.3 | NP_476494.2 | 7 | 1394 | |||
| GenBank mRNA | BC166763.1 | - | 7 | 1610 | 7931847 | |
| DY318631.1 | - | 1 | 529 | |||

Oxr1
Genomic Map