siRNA Details
| Gene Symbol: Ep300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765878.1 | XP_008764100.1 | 2 | 1308 | ||
| GenBank mRNA | AB066220.1 | - | 2 | 338 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ep300| Gene Symbol: Ep300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765878.1 | XP_008764100.1 | 2 | 1308 | ||
| GenBank mRNA | AB066220.1 | - | 2 | 338 | ||