siRNA Details
| Gene Symbol: Pank4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133531.2 | NP_598215.1 | 3, 4 | 428 | ||
| GenBank mRNA | AF399873.1 | - | 3, 4 | 428 | ||
| FQ224186.1 | - | 3, 4 | 432 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pank4| Gene Symbol: Pank4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133531.2 | NP_598215.1 | 3, 4 | 428 | ||
| GenBank mRNA | AF399873.1 | - | 3, 4 | 428 | ||
| FQ224186.1 | - | 3, 4 | 432 | |||