siRNA Details
| Gene Symbol: Mrpl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133539.1 | NP_598223.1 | 3 | 497 | ||
| GenBank mRNA | U53512.1 | - | 3 | 497 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrpl17| Gene Symbol: Mrpl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133539.1 | NP_598223.1 | 3 | 497 | ||
| GenBank mRNA | U53512.1 | - | 3 | 497 | ||