siRNA Details
| Gene Symbol: Timm9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276429.1 | NP_001263358.1 | 4 | 296 | ||
| NM_001276430.1 | NP_001263359.1 | 3 | 281 | |||
| NM_133604.1 | NP_598288.1 | 4 | 367 | |||
| GenBank mRNA | CB775939.1 | - | 4 | 346 | ||
| FM052814.1 | - | 4 | 205 | |||
| FM056491.1 | - | 3 | 244 | |||
| FQ228524.1 | - | 4 | 367 | |||

Timm9
Genomic Map