siRNA Details
| Gene Symbol: Mat2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134351.1 | NP_599178.1 | 5 | 616 | ||
| GenBank mRNA | BC062394.1 | - | 649 | 5622019 | ||
| J05571.1 | - | 5 | 616 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mat2a| Gene Symbol: Mat2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134351.1 | NP_599178.1 | 5 | 616 | ||
| GenBank mRNA | BC062394.1 | - | 649 | 5622019 | ||
| J05571.1 | - | 5 | 616 | |||