siRNA Details
Gene Symbol: Mat2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134351.1 | NP_599178.1 | 5 | 527 | ||
GenBank mRNA | BC062394.1 | - | 560 | 5622019 | ||
J05571.1 | - | 5 | 527 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mat2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134351.1 | NP_599178.1 | 5 | 527 | ||
GenBank mRNA | BC062394.1 | - | 560 | 5622019 | ||
J05571.1 | - | 5 | 527 |