siRNA Details
| Gene Symbol: Trpm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134371.2 | NP_599198.2 | 6 | 613 | ||
| GenBank mRNA | AY072788.2 | - | 6 | 613 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trpm8| Gene Symbol: Trpm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134371.2 | NP_599198.2 | 6 | 613 | ||
| GenBank mRNA | AY072788.2 | - | 6 | 613 | ||