siRNA Details
| Gene Symbol: Lphn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF063102.1 | - | 23, 24 | 3740 | ||
| AF081148.1 | - | 22, 23 | 4081 | |||
| AF081150.1 | - | 22, 23 | 4114 | |||
| AF081151.1 | - | 23, 24 | 4126 | |||
| AF081153.1 | - | 23, 24 | 4159 | |||

Lphn2
Genomic Map