siRNA Details
| Gene Symbol: Phf5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138888.1 | NP_620243.1 | 4 | 754 | ||
| GenBank mRNA | AF495522.1 | - | 4 | 754 | ||
| BC168144.1 | - | 4 | 772 | 8372591 | ||
| FQ223455.1 | - | 2 | 528 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phf5a| Gene Symbol: Phf5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138888.1 | NP_620243.1 | 4 | 754 | ||
| GenBank mRNA | AF495522.1 | - | 4 | 754 | ||
| BC168144.1 | - | 4 | 772 | 8372591 | ||
| FQ223455.1 | - | 2 | 528 | |||