siRNA Details
Gene Symbol: Dctpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138892.1 | NP_620247.1 | 3 | 632 | ||
GenBank mRNA | AY029335.1 | - | 3 | 632 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dctpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138892.1 | NP_620247.1 | 3 | 632 | ||
GenBank mRNA | AY029335.1 | - | 3 | 632 |