siRNA Details
| Gene Symbol: Gls2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270786.1 | NP_001257715.1 | 14 | 1645 | ||
| NM_001270787.1 | NP_001257716.1 | 14 | 1891 | |||
| NM_138904.2 | NP_620259.2 | 13 | 1340 | |||
| XM_006240730.2 | XP_006240792.1 | 15 | 1470 | |||
| GenBank mRNA | BC089776.1 | - | 14 | 1484 | ||
| BC104712.1 | - | 14 | 1389 | |||
| FQ211735.1 | - | 14 | 1891 | |||
| J05499.1 | - | 13 | 1339 | |||

Gls2
Genomic Map