siRNA Details
| Gene Symbol: Coro7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191639.1 | NP_001178568.1 | 278 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Coro7| Gene Symbol: Coro7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191639.1 | NP_001178568.1 | 278 | |||