siRNA Details
| Gene Symbol: Fnbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138914.1 | NP_620269.1 | 427 | |||
| XM_006233929.2 | XP_006233991.1 | 6 | 1823 | |||
| GenBank mRNA | AB073208.1 | - | 427 | |||
| AB126168.1 | - | 427 | ||||
| AB126169.1 | - | 427 | ||||
| AB126170.1 | - | 427 | ||||
| AB126171.1 | - | 427 | ||||
| AB126172.1 | - | 427 | ||||

Fnbp1
Genomic Map