siRNA Details
| Gene Symbol: LOC100910472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241915.2 | XP_006241977.1 | 2 | 797 | ||
| XM_006241916.2 | XP_006241978.1 | 2 | 813 | |||
| XM_006241917.2 | XP_006241979.1 | 2 | 787 | |||
| XM_006241918.2 | XP_006241980.1 | 2 | 775 | |||
| XM_008765634.1 | XP_008763856.1 | 2 | 818 | |||
| Gene Symbol: Commd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139108.2 | NP_620808.1 | 484 | |||
| XM_006241924.2 | XP_006241986.1 | 2 | 797 | |||
| XM_006241925.2 | XP_006241987.1 | 2 | 787 | |||
| XM_006241926.2 | XP_006241988.1 | 2 | 775 | |||
| XM_008765544.1 | XP_008763766.1 | 2 | 818 | |||
| GenBank mRNA | AF290194.1 | - | 489 | |||

LOC100910472
Genomic Map