siRNA Details
| Gene Symbol: Rpl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139114.3 | NP_620814.1 | 5 | 1068 | ||
| GenBank mRNA | BC078724.1 | - | 4 | 1007 | 7125608 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpl15| Gene Symbol: Rpl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139114.3 | NP_620814.1 | 5 | 1068 | ||
| GenBank mRNA | BC078724.1 | - | 4 | 1007 | 7125608 | |