siRNA Details
| Gene Symbol: Uxs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139336.1 | NP_647552.1 | 15 | 1194 | ||
| XM_006244296.2 | XP_006244358.1 | 15 | 1252 | |||
| XM_008766751.1 | XP_008764973.1 | 16 | 1710 | |||
| GenBank mRNA | AF482705.1 | - | 15 | 1194 | ||
| BC086988.1 | - | 15 | 1205 | 7192831 | ||

Uxs1
Genomic Map