siRNA Details
| Gene Symbol: Uxs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139336.1 | NP_647552.1 | 4 | 232 | ||
| XM_006244296.2 | XP_006244358.1 | 4 | 290 | |||
| XM_008766746.1 | XP_008764968.1 | 5 | 748 | |||
| XM_008766747.1 | XP_008764969.1 | 400 | ||||
| XM_008766748.1 | XP_008764970.1 | 5 | 733 | |||
| XM_008766749.1 | XP_008764971.1 | 4 | 292 | |||
| XM_008766750.1 | XP_008764972.1 | 4 | 277 | |||
| XM_008766751.1 | XP_008764973.1 | 5 | 748 | |||
| XM_008766752.1 | XP_008764974.1 | 4 | 640 | |||
| XM_008766753.1 | XP_008764975.1 | 4 | 625 | |||
| GenBank mRNA | AF482705.1 | - | 4 | 232 | ||
| BC086988.1 | - | 4 | 243 | 7192831 | ||

Uxs1
Genomic Map