siRNA Details
| Gene Symbol: Gpsm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145469.1 | NP_001138941.1 | 2 | 195 | ||
| NM_144745.2 | NP_653346.2 | 12 | 1635 | |||
| XM_006233670.2 | XP_006233732.1 | 12 | 1739 | |||
| XM_008761620.1 | XP_008759842.1 | 4 | 3495 | |||
| GenBank mRNA | AF107723.1 | - | 11 | 1759 | ||
| BC086535.1 | - | 2 | 195 | |||

Gpsm1
Genomic Map