siRNA Details
| Gene Symbol: Pdp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145091.4 | NP_659559.2 | 2 | 717 | ||
| XM_006255048.2 | XP_006255110.1 | 2 | 762 | |||
| GenBank mRNA | AF062741.1 | - | 2 | 617 | ||
| BC072485.1 | - | 717 | 7105072 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdp2| Gene Symbol: Pdp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145091.4 | NP_659559.2 | 2 | 717 | ||
| XM_006255048.2 | XP_006255110.1 | 2 | 762 | |||
| GenBank mRNA | AF062741.1 | - | 2 | 617 | ||
| BC072485.1 | - | 717 | 7105072 | |||