siRNA Details
| Gene Symbol: Cby1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145676.1 | NP_663709.1 | 4, 5 | 390 | ||
| XM_006241967.2 | XP_006242029.1 | 5, 6 | 507 | |||
| GenBank mRNA | AF393211.1 | - | 4, 5 | 390 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cby1| Gene Symbol: Cby1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145676.1 | NP_663709.1 | 4, 5 | 390 | ||
| XM_006241967.2 | XP_006242029.1 | 5, 6 | 507 | |||
| GenBank mRNA | AF393211.1 | - | 4, 5 | 390 | ||