siRNA Details
| Gene Symbol: Eln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012722.1 | NP_036854.1 | 37 | 2919 | ||
| GenBank mRNA | BC085910.1 | - | 37 | 2919 | 7097661 | |
| J04035.1 | - | 6 | 541 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eln| Gene Symbol: Eln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012722.1 | NP_036854.1 | 37 | 2919 | ||
| GenBank mRNA | BC085910.1 | - | 37 | 2919 | 7097661 | |
| J04035.1 | - | 6 | 541 | |||