siRNA Details
| Gene Symbol: Bles03 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024233.2 | NP_001019404.2 | 2 | 900 | ||
| NM_001243534.1 | NP_001230463.1 | 2 | 914 | |||
| GenBank mRNA | AF502571.2 | - | 1 | 107 | ||
| BC093387.1 | - | 2 | 759 | 7389419 | ||
| FQ233132.1 | - | 2 | 793 | |||

Bles03
Genomic Map