siRNA Details
| Gene Symbol: Gzma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153468.1 | NP_703198.1 | 2 | 142 | ||
| GenBank mRNA | AB082125.1 | - | 2 | 142 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gzma| Gene Symbol: Gzma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153468.1 | NP_703198.1 | 2 | 142 | ||
| GenBank mRNA | AB082125.1 | - | 2 | 142 | ||