siRNA Details
| Gene Symbol: Vom2r40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173317.1 | NP_775439.1 | 2567 | |||
| GenBank mRNA | AF016180.1 | - | 2567 | |||
| Gene Symbol: Vom2r41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099500.1 | NP_001092970.1 | 6 | 1927 | ||
More Information
Set Membership: |
|

Vom2r40
Genomic Map