siRNA Details
| Gene Symbol: Akap17a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127246.1 | NP_001120718.1 | 5 | 1685 | ||
| XM_006248998.2 | XP_006249060.1 | 5 | 2398 | |||
| XM_006248999.2 | XP_006249061.1 | 5 | 2242 | |||
| XM_006249000.2 | XP_006249062.1 | 5 | 2333 | |||
| GenBank mRNA | BC100662.1 | - | 5 | 1641 | ||
| BC166722.1 | - | 5 | 1685 | 7933458 | ||

Akap17a
Genomic Map