siRNA Details
| Gene Symbol: Cc2d1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013869.1 | NP_001013891.1 | 20 | 2208 | ||
| XM_006255233.2 | XP_006255295.1 | 21 | 2400 | |||
| XM_006255234.2 | XP_006255296.1 | 21 | 2397 | |||
| XM_008772364.1 | XP_008770586.1 | 20 | 2266 | |||
| GenBank mRNA | BC081948.1 | - | 20 | 2208 | 7122187 | |

Cc2d1a
Genomic Map