siRNA Details
Gene Symbol: LOC288913 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198728.1 | NP_942023.1 | 3 | 331 | ||
GenBank mRNA | X62277.1 | - | 3 | 331 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC288913 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198728.1 | NP_942023.1 | 3 | 331 | ||
GenBank mRNA | X62277.1 | - | 3 | 331 |