siRNA Details
| Gene Symbol: LOC288913 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198728.1 | NP_942023.1 | 3 | 484 | ||
| GenBank mRNA | X62277.1 | - | 3 | 484 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC288913| Gene Symbol: LOC288913 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198728.1 | NP_942023.1 | 3 | 484 | ||
| GenBank mRNA | X62277.1 | - | 3 | 484 | ||